Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Psmb6
Entrez Gene ID
19175
Gene Name
proteasome (prosome, macropain) subunit, beta type 6
Gene Aliases
Lmp19, Mpnd
Chromosome Location
Chr.11: 70525357 - 70527858 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008946.4 NP_032972.3 2 - 3 209 60
GenBank mRNA AK078437.1 - 2 - 3 181 60
AK132042.1 - 2 - 3 192 60
AK167123.1 - 2 - 3 181 60
AK167227.1 - 2 - 3 177 60
BC013897.1 - 2 - 3 169 60
BY069663.1 - 2 - 3 209 60
U13393.1 - 2 - 3 130 60

View Full Product Details