The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010157.3 | NP_034287.3 | 8 - 9 | 1808 | 69 |
NM_207707.1 | NP_997590.1 | 9 - 10 | 1862 | 69 | |
NR_104386.1 | 7 - 8 | 1669 | 69 | ||
GenBank mRNA | AK054290.1 | - | 9 - 10 | 1860 | 69 |
AK054301.1 | - | 7 - 8 | 1670 | 69 | |
AK054413.1 | - | 9 - 10 | 1863 | 69 | |
AY054413.1 | - | 8 - 9 | 1323 | 69 | |
BC141075.1 | - | 7 - 8 | 1510 | 69 | |
BC145329.1 | - | 7 - 8 | 1510 | 69 | |
U81451.1 | - | 7 - 8 | 1280 | 69 |