Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rph3a
Entrez Gene ID
19894
Gene Name
rabphilin 3A
Gene Aliases
2900002P20Rik, AU022689, AW108370
Chromosome Location
Chr.5: 120940499 - 121010102 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB112924.1 - 8 - 9 845 101
AK139040.1 - 7 - 8 787 101
BC042585.1 - 8 - 9 1012 101
BC050883.1 - 8 - 9 979 101
D29965.1 - 8 - 9 822 101

View Full Product Details