Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Spag1
Entrez Gene ID
26942
Gene Name
sperm associated antigen 1
Gene Aliases
tpis
Chromosome Location
Chr.15: 36175483 - 36235621 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012031.3 NP_036161.2 11 - 12 1623 65
GenBank mRNA AF181252.1 - 11 - 12 1615 65
AF181253.1 - 1 - 2 307 65
AK032601.1 - 8 - 9 1418 65
BC046313.1 - 11 - 12 1623 65

View Full Product Details