The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012034.3 | NP_036164.2 | 2 - 3 | 324 | 78 |
GenBank mRNA | AF175523.1 | - | 2 - 3 | 225 | 78 |
BC019974.1 | - | 2 - 3 | 248 | 78 | |
BC052325.1 | - | 2 - 3 | 293 | 78 |