The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281854.1 | NP_001268783.1 | 1 - 2 | 406 | 65 |
NM_012054.4 | NP_036184.1 | 1 - 2 | 406 | 65 | |
GenBank mRNA | AK084452.1 | - | 1 - 2 | 401 | 65 |
AK085343.1 | - | 1 - 2 | 376 | 65 | |
AK133301.1 | - | 1 - 2 | 347 | 65 | |
AK150886.1 | - | 1 - 2 | 382 | 65 | |
AK152346.1 | - | 1 - 2 | 382 | 65 | |
AK171572.1 | - | 1 - 2 | 225 | 65 | |
BC089580.1 | - | 1 - 2 | 386 | 65 | |
BC150767.1 | - | 1 - 2 | 200 | 65 | |
CT010242.1 | - | 1 - 2 | 125 | 65 |