The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013879.2 | NP_038907.1 | 2 - 3 | 290 | 66 |
GenBank mRNA | AF169152.1 | - | 2 - 3 | 290 | 66 |
AK141231.1 | - | 2 - 3 | 204 | 66 |