The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013891.4 | NP_038919.1 | 2 - 3 | 831 | 67 |
GenBank mRNA | AB019436.1 | - | 2 - 3 | 767 | 67 |
AK078863.1 | - | 2 - 3 | 832 | 67 | |
BC012648.1 | - | 2 - 3 | 832 | 67 |