Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mmp20
Entrez Gene ID
30800
Gene Name
matrix metallopeptidase 20 (enamelysin)
Gene Aliases
-
Chromosome Location
Chr.9: 7628231 - 7674968 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013903.2 NP_038931.1 7 - 8 1110 69
GenBank mRNA AF155933.1 - 7 - 8 1110 69
BC152335.1 - 7 - 8 1114 69
BC152336.1 - 7 - 8 1114 69

View Full Product Details