Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sart1
Entrez Gene ID
20227
Gene Name
squamous cell carcinoma antigen recognized by T cells 1
Gene Aliases
U5-110K
Chromosome Location
Chr.19: 5377523 - 5389420 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016882.3 NP_058578.3 11 - 12 1519 64
GenBank mRNA AB014721.1 - 11 - 12 1477 64
AF129931.1 - 11 - 12 1519 64
AK088341.1 - 11 - 12 1480 64
BC028823.1 - 11 - 12 1481 64
BC051394.1 - 11 - 12 1481 64

View Full Product Details