Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sart1
Entrez Gene ID
20227
Gene Name
squamous cell carcinoma antigen recognized by T cells 1
Gene Aliases
U5-110K
Chromosome Location
Chr.19: 5377523 - 5389420 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016882.3 NP_058578.3 7 - 8 917 80
GenBank mRNA AB014721.1 - 7 - 8 875 80
AF129931.1 - 7 - 8 917 80
AK088341.1 - 7 - 8 877 80
BC028823.1 - 7 - 8 879 80
BC051394.1 - 7 - 8 879 80

View Full Product Details