Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Osbpl5
Entrez Gene ID
79196
Gene Name
oxysterol binding protein-like 5
Gene Aliases
1110006M06Rik, AI462538, ORP5, Obph1, Osbp2
Chromosome Location
Chr.7: 143688762 - 143756980 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199227.1 NP_001186156.1 13 - 14 1634 65
NM_024289.2 NP_077251.1 13 - 14 1834 65
GenBank mRNA AB074008.1 - 13 - 14 1627 65
AJ278263.2 - 13 - 14 1834 65
AK129385.1 - 4 - 5 590 65
AK184679.1 - 2 - 3 136 65
BC079872.1 - 13 - 14 1615 65

View Full Product Details