The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290518.1 | NP_001277447.1 | 2 - 3 | 181 | 80 |
NM_025286.3 | NP_079562.1 | 2 - 3 | 219 | 80 | |
GenBank mRNA | AK003269.1 | - | 2 - 3 | 154 | 80 |
AK007579.1 | - | 2 - 3 | 176 | 80 | |
AK008473.1 | - | 2 - 3 | 203 | 80 | |
AK015878.1 | - | 2 - 3 | 182 | 80 | |
AK152812.1 | - | 2 - 3 | 176 | 80 | |
AK188325.1 | - | 1 - 2 | 64 | 80 | |
BB853212.1 | - | 2 - 3 | 219 | 80 | |
BC029183.1 | - | 2 - 3 | 110 | 80 | |
BC099447.1 | - | 2 - 3 | 185 | 80 |