Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nfkbiz
Entrez Gene ID
80859
Gene Name
nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor, zeta
Gene Aliases
AA408868, INAP, Mail
Chromosome Location
Chr.16: 55811377 - 55838641 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159394.1 NP_001152866.1 1 - 2 374 147
NM_030612.3 NP_085115.1 3 - 4 604 147
GenBank mRNA AB020974.1 - 3 - 4 816 147
AB026551.2 - 1 - 2 374 147
AK082908.1 - 1 - 2 373 147
AK141318.1 - 1 - 2 373 147
AK150519.1 - 1 - 2 373 147
AK152016.1 - 1 - 2 375 147
AK156826.1 - 1 - 2 375 147
AK159093.1 - 1 - 2 375 147
AK159171.1 - 1 - 2 375 147
AK159459.1 - 1 - 2 375 147
AK159645.1 - 1 - 2 375 147
AK159917.1 - 1 - 2 375 147
AK160065.1 - 1 - 2 373 147
BC058188.1 - 1 - 2 359 147

View Full Product Details