The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080728.2 | NP_542766.1 | 40 - 41 | 5926 | 102 |
GenBank mRNA | AJ223362.1 | - | 1 - 2 | 73 | 102 |
AK147031.1 | - | 40 - 41 | 5927 | 102 | |
AK168593.1 | - | 21 - 22 | 3695 | 102 | |
AK168677.1 | - | 24 - 25 | 3957 | 102 | |
AK169198.1 | - | 40 - 41 | 5926 | 102 | |
AK169371.1 | - | 40 - 41 | 5926 | 102 | |
AY056464.1 | - | 40 - 41 | 5926 | 102 | |
BC121789.1 | - | 39 - 40 | 5882 | 102 | |
M74752.1 | - | 15 - 16 | 2581 | 102 |