Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bpifb1
Entrez Gene ID
228801
Gene Name
BPI fold containing family B, member 1
Gene Aliases
Lplunc1
Chromosome Location
Chr.2: 154190818 - 154220343 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012392.1 NP_001012392.1 11 - 12 1258 73
NM_153418.2 NP_700467.2 11 - 12 1221 73
GenBank mRNA AK146605.1 - 11 - 12 1254 73
BC060982.1 - 11 - 12 1258 73
U46068.3 - 11 - 12 1222 73

View Full Product Details