Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bpifb1
Entrez Gene ID
228801
Gene Name
BPI fold containing family B, member 1
Gene Aliases
Lplunc1
Chromosome Location
Chr.2: 154190818 - 154220343 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012392.1 NP_001012392.1 5 - 6 630 102
NM_153418.2 NP_700467.2 5 - 6 593 102
GenBank mRNA AK146605.1 - 5 - 6 626 102
BC060982.1 - 5 - 6 630 102
BC089529.1 - - 630 102
BC100436.1 - 5 - 6 625 102
U46068.3 - 5 - 6 594 102

View Full Product Details