The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013902.4 | NP_038930.1 | 3 - 4 | 337 | 147 |
GenBank mRNA | AF110812.1 | - | 3 - 4 | 334 | 147 |
AF135595.1 | - | 3 - 4 | 339 | 147 | |
AK012346.1 | - | 3 - 4 | 332 | 147 | |
AK145648.1 | - | 3 - 4 | 345 | 147 | |
AK150802.1 | - | 3 - 4 | 347 | 147 | |
AK168641.1 | - | 3 - 4 | 344 | 147 | |
BC002122.1 | - | 3 - 4 | 361 | 147 |