The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080795.4 | NP_542985.4 | 2 - 3 | 625 | 128 |
GenBank mRNA | AF401681.1 | - | 2 - 3 | 582 | 128 |
AK036218.1 | - | 2 - 3 | 626 | 128 | |
AK047053.1 | - | 2 - 3 | 609 | 128 | |
AK140320.1 | - | 2 - 3 | 559 | 128 | |
AK170146.1 | - | 2 - 3 | 579 | 128 | |
BC090665.1 | - | 2 - 3 | 591 | 128 | |
BC117937.1 | - | 2 - 3 | 580 | 128 | |
BC117938.1 | - | 2 - 3 | 579 | 128 |