Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ahnak
Entrez Gene ID
66395
Gene Name
AHNAK nucleoprotein (desmoyokin)
Gene Aliases
DY6
Chromosome Location
Chr.19: 8989125 - 9076935 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_104460.1 5 - 6 702 80
GenBank mRNA AK009866.1 - 5 - 6 725 80

View Full Product Details