Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trak1
Entrez Gene ID
67095
Gene Name
trafficking protein, kinesin binding 1
Gene Aliases
2310001H13Rik, AI413908, AI467545, hyrt, mKIAA1042
Chromosome Location
Chr.9: 121297507 - 121474919 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175114.3 NP_780323.2 12 - 13 1569 85
GenBank mRNA AK029123.1 - 13 - 14 1967 85
AK041436.1 - 13 - 14 1937 85
AK122426.1 - 13 - 14 1659 85
BC058971.1 - 12 - 13 1511 85
BC116262.1 - 12 - 13 1536 85
BC116263.1 - 12 - 13 1536 85
BC129961.1 - 12 - 13 1536 85
BC129962.1 - 12 - 13 1536 85

View Full Product Details