The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175130.4 | NP_780339.2 | 9 - 10 | 1225 | 78 |
GenBank mRNA | AJ575814.1 | - | 9 - 10 | 1204 | 78 |
AK044094.1 | - | - | 832 | 78 | |
BC049165.1 | - | 7 - 8 | 856 | 78 | |
BC058632.1 | - | 2 - 3 | 133 | 78 | |
BC096475.1 | - | 9 - 10 | 1201 | 78 |