Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dnaic1
Entrez Gene ID
68922
Gene Name
dynein, axonemal, intermediate chain 1
Gene Aliases
1110066F04Rik, BB124644, Dnai1, b2b1526Clo
Chromosome Location
Chr.4: 41569663 - 41638158 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175138.4 NP_780347.2 14 - 15 1603 79
GenBank mRNA AK030213.1 - 14 - 15 1537 79
AK047808.1 - 14 - 15 1604 79
BC138785.1 - 14 - 15 1548 79
BC138787.1 - 14 - 15 1548 79

View Full Product Details