The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175138.4 | NP_780347.2 | 18 - 19 | 2018 | 83 |
GenBank mRNA | AK030213.1 | - | 18 - 19 | 1952 | 83 |
AK047808.1 | - | 18 - 19 | 2019 | 83 | |
BC138785.1 | - | 18 - 19 | 1963 | 83 | |
BC138787.1 | - | 18 - 19 | 1963 | 83 | |
CD775688.1 | - | 2 - 3 | 83 |