The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175151.4 | NP_780360.1 | 10 - 11 | 699 | 139 |
GenBank mRNA | AK010232.1 | - | 8 - 9 | 707 | 139 |
AK042248.1 | - | 10 - 11 | 700 | 139 | |
BC034895.1 | - | 7 - 8 | 486 | 139 | |
BC061187.1 | - | 10 - 11 | 696 | 139 |