Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tatdn1
Entrez Gene ID
69694
Gene Name
TatD DNase domain containing 1
Gene Aliases
2310079P03Rik, CDA11
Chromosome Location
Chr.15: 58890153 - 58933781 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175151.4 NP_780360.1 4 - 5 225 124
GenBank mRNA AK042248.1 - 4 - 5 226 124
BC061187.1 - 4 - 5 222 124

View Full Product Details