The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175164.4 | NP_780373.3 | 21 - 22 | 2599 | 65 |
GenBank mRNA | AK129176.1 | - | 21 - 22 | 2537 | 65 |