Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Golim4
Entrez Gene ID
73124
Gene Name
golgi integral membrane protein 4
Gene Aliases
3110027H23Rik, GIMPC, GPP130, Golph4, P138
Chromosome Location
Chr.3: 75876183 - 75956949 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291069.1 NP_001277998.1 3 - 4 966 147
NM_175193.6 NP_780402.1 3 - 4 966 147
GenBank mRNA AK033730.1 - 3 - 4 953 147
AK048434.1 - 4 - 5 877 147
AK078928.1 - 3 - 4 966 147
AK149286.1 - 3 - 4 926 147
BC086642.1 - 3 - 4 798 147
BC089354.1 - 3 - 4 848 147

View Full Product Details