Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tasp1
Entrez Gene ID
75812
Gene Name
taspase, threonine aspartase 1
Gene Aliases
4930485D02Rik, AW986064
Chromosome Location
Chr.2: 139833479 - 140067666 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159640.2 NP_001153112.1 5 - 6 522 64
NM_001159641.2 NP_001153113.1 5 - 6 522 64
NM_001289611.1 NP_001276540.1 5 - 6 522 64
NM_175225.5 NP_780434.1 5 - 6 789 64
NR_110350.1 5 - 6 789 64
NR_110351.1 5 - 6 522 64
NR_110352.1 4 - 5 453 64
GenBank mRNA AK015624.1 - 5 - 6 522 64
AK036512.1 - 5 - 6 745 64
AK050603.1 - 2 - 3 246 64
AK132709.1 - 4 - 5 439 64
AK146320.1 - 5 - 6 505 64
AK160983.1 - 5 - 6 750 64
BB665115.1 - 5 - 6 498 64
BC024597.1 - 5 - 6 509 64

View Full Product Details