The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252494.1 | NP_001239423.1 | 9 - 10 | 1149 | 122 |
NM_001252496.1 | NP_001239425.1 | 9 - 10 | 1149 | 122 | |
NM_001252497.1 | NP_001239426.1 | 4 - 5 | 1407 | 122 | |
NM_001252498.1 | NP_001239427.1 | 4 - 5 | 1407 | 122 | |
NM_175258.4 | NP_780467.2 | 9 - 10 | 1149 | 122 | |
GenBank mRNA | AK045540.1 | - | 9 - 10 | 1091 | 122 |
AK049228.1 | - | 9 - 10 | 1105 | 122 | |
AK049669.1 | - | 4 - 5 | 1407 | 122 | |
AK164343.1 | - | 4 - 5 | 1380 | 122 | |
AK167055.1 | - | 9 - 10 | 1150 | 122 | |
BC141182.1 | - | 9 - 10 | 1048 | 122 | |
BC145503.1 | - | 9 - 10 | 1048 | 122 |