The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175300.4 | NP_780509.2 | 10 - 11 | 1980 | 69 |
GenBank mRNA | AA727217.1 | - | 3 - 4 | 308 | 69 |
AK033784.1 | - | 10 - 11 | 1948 | 69 | |
AK129351.1 | - | 10 - 11 | 1933 | 69 | |
AK154465.1 | - | 10 - 11 | 1934 | 69 | |
AK170676.1 | - | 10 - 11 | 1946 | 69 | |
BC027351.1 | - | 10 - 11 | 1928 | 69 | |
BC065421.1 | - | 1 - 2 | 93 | 69 | |
BC096381.1 | - | 9 - 10 | 2173 | 69 | |
BC137583.1 | - | 10 - 11 | 1948 | 69 |