The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175314.3 | NP_780523.2 | 19 - 20 | 2868 | 92 |
GenBank mRNA | AK030699.1 | - | 6 - 7 | 978 | 92 |
AK078802.1 | - | 1 - 2 | 63 | 92 | |
BC068142.1 | - | 5 - 6 | 907 | 92 |