Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bbs4
Entrez Gene ID
102774
Gene Name
Bardet-Biedl syndrome 4 (human)
Gene Aliases
AW537059, AW742241, D9Ertd464e
Chromosome Location
Chr.9: 59321966 - 59353508 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175325.3 NP_780534.1 7 - 8 536 81
GenBank mRNA AK039560.1 - 7 - 8 485 81
AK089970.1 - 7 - 8 491 81
AK134734.1 - 7 - 8 489 81
BC055797.1 - 7 - 8 456 81
BC089507.1 - 7 - 8 470 81
BC092531.1 - 7 - 8 480 81
BC145771.1 - 7 - 8 498 81
BC145773.1 - 7 - 8 498 81

View Full Product Details