The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253813.1 | NP_001240742.1 | 9 - 10 | 1205 | 76 |
NM_175344.4 | NP_780553.2 | 8 - 9 | 1142 | 76 | |
GenBank mRNA | AK089300.1 | - | 8 - 9 | 1055 | 76 |
AK159411.1 | - | 9 - 10 | 1154 | 76 | |
BC060732.1 | - | 8 - 9 | 1142 | 76 |