Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zdhhc15
Entrez Gene ID
108672
Gene Name
zinc finger, DHHC domain containing 15
Gene Aliases
6030457O13Rik
Chromosome Location
Chr.X: 104536969 - 104671090 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175358.4 NP_780567.1 3 - 4 520 138
GenBank mRNA AK029137.1 - 3 - 4 260 138
AK032922.1 - 3 - 4 241 138
AK077949.1 - 3 - 4 269 138
AK132304.1 - 3 - 4 521 138
AK160360.1 - 3 - 4 260 138

View Full Product Details