The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175483.5 | NP_780692.2 | 1 - 2 | 3060 | 67 |
GenBank mRNA | AK048566.1 | - | 1 - 2 | 1565 | 67 |
AK053761.1 | - | 1 - 2 | 1595 | 67 | |
AK142274.1 | - | 1 - 2 | 3061 | 67 | |
BC096472.1 | - | 1 - 2 | 1810 | 67 |