Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Plch2
Entrez Gene ID
269615
Gene Name
phospholipase C, eta 2
Gene Aliases
A930027K05Rik, PLCeta2, Plc-eta2, Plcl4
Chromosome Location
Chr.4: 154983115 - 155064091 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113360.2 NP_001106831.1 5 - 6 1041 85
NM_175556.4 NP_780765.2 5 - 6 1041 85
GenBank mRNA AK044619.1 - 5 - 6 1040 85
AK078731.1 - 5 - 6 812 85
AK129147.1 - 5 - 6 741 85
AY966876.1 - 5 - 6 819 85
DQ176851.1 - 5 - 6 1041 85

View Full Product Details