The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175562.3 | NP_780771.1 | 1 - 2 | 336 | 87 |
GenBank mRNA | AB232635.1 | - | 1 - 2 | 219 | 87 |
AK049038.1 | - | 1 - 2 | 336 | 87 | |
AK053487.1 | - | 1 - 2 | 333 | 87 | |
BC049787.1 | - | 1 - 2 | 316 | 87 |