The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_176840.3 | NP_789810.2 | 6 - 7 | 1633 | 107 |
GenBank mRNA | BC035278.1 | - | 6 - 7 | 1633 | 107 |