The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164531.1 | NP_001158003.1 | 3 - 4 | 371 | 66 |
NM_177319.3 | NP_796293.2 | 3 - 4 | 488 | 66 | |
GenBank mRNA | AK159065.1 | - | 3 - 4 | 345 | 66 |
BC024801.1 | - | 3 - 4 | 462 | 66 | |
BC042595.1 | - | 3 - 4 | 434 | 66 | |
CK333536.1 | - | 3 - 4 | 339 | 66 | |
CK781433.1 | - | 3 - 4 | 488 | 66 |