The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177388.3 | NP_796362.1 | 2 - 3 | 798 | 115 |
GenBank mRNA | AK038549.1 | - | 2 - 3 | 799 | 115 |
BC138146.1 | - | 1 - 2 | 636 | 115 | |
BC138147.1 | - | 1 - 2 | 636 | 115 |