The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040398.2 | NP_001035488.2 | 16 - 17 | 6061 | 69 |
GenBank mRNA | AK122435.1 | - | 8 - 9 | 2396 | 69 |
BC023450.1 | - | 2 - 3 | 242 | 69 | |
BC035291.1 | - | 8 - 9 | 2563 | 69 | |
BC038367.1 | - | 10 - 11 | 3139 | 69 | |
BC040775.1 | - | 8 - 9 | 2563 | 69 | |
BC041681.1 | - | 8 - 9 | 2582 | 69 |