The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177595.4 | NP_808263.2 | 6 - 7 | 1115 | 61 |
GenBank mRNA | AK034675.1 | - | 6 - 7 | 1093 | 61 |
BC137728.1 | - | 6 - 7 | 1050 | 61 | |
CD544112.1 | - | 2 - 3 | 96 | 61 |