Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pstk
Entrez Gene ID
214580
Gene Name
phosphoseryl-tRNA kinase
Gene Aliases
5430423O14Rik, 5730458D16Rik, Gm1099, R75284
Chromosome Location
Chr.7: 131371113 - 131392768 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK077585.1 - 4 - 5 806 109

View Full Product Details