Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Os9
Entrez Gene ID
216440
Gene Name
amplified in osteosarcoma
Gene Aliases
4632413K17Rik, AU022351
Chromosome Location
Chr.10: 127094259 - 127121160 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171026.1 NP_001164497.1 4 - 5 521 57
NM_177614.3 NP_808282.2 4 - 5 521 57
GenBank mRNA AK028047.1 - 4 - 5 496 57
BC006844.1 - 4 - 5 341 57
BC031768.1 - 4 - 5 486 57
BU703218.1 - 4 - 5 485 57

View Full Product Details