The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178060.3 | NP_835161.1 | 2 - 3 | 484 | 82 |
GenBank mRNA | AK078233.1 | - | 2 - 3 | 484 | 82 |
BC046795.1 | - | - | 456 | 82 | |
X07750.1 | - | 1 - 2 | 54 | 82 | |
X07751.1 | - | - | 54 | 82 | |
X07752.1 | - | - | 54 | 82 | |
X51983.1 | - | 1 - 2 | 58 | 82 |