Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Agfg2
Entrez Gene ID
231801
Gene Name
ArfGAP with FG repeats 2
Gene Aliases
A630095P14Rik, Hrbl, RAB-R, RABR
Chromosome Location
Chr.5: 137650463 - 137684806 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK042486.1 - 6 - 7 904 72
AK050881.1 - 7 - 8 651 72
AK155221.1 - 12 - 13 1524 72
BC046788.1 - 11 - 12 1427 72

View Full Product Details