The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177231.2 | NP_796205.1 | 14 - 15 | 1388 | 97 |
NM_178220.3 | NP_835738.1 | 13 - 14 | 1364 | 97 | |
GenBank mRNA | AK036421.1 | - | 3 - 4 | 295 | 97 |
AK052588.1 | - | 14 - 15 | 1165 | 97 | |
AK090090.1 | - | 13 - 14 | 1191 | 97 | |
AK144054.1 | - | 14 - 15 | 1187 | 97 | |
AK147508.1 | - | 14 - 15 | 1389 | 97 | |
AK147566.1 | - | 14 - 15 | 1410 | 97 | |
AK156213.1 | - | 3 - 4 | 298 | 97 | |
AK165514.1 | - | 14 - 15 | 1187 | 97 | |
BC108969.1 | - | 13 - 14 | 1131 | 97 | |
BC108970.1 | - | 13 - 14 | 1131 | 97 |