Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Reps2
Entrez Gene ID
194590
Gene Name
RALBP1 associated Eps domain containing protein 2
Gene Aliases
POB1
Chromosome Location
Chr.X: 162411952 - 162643705 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290633.1 NP_001277562.1 15 - 16 1776 75
NM_178256.4 NP_839987.2 15 - 16 1779 75
GenBank mRNA AK147418.1 - 13 - 14 1241 75
AK147432.1 - 12 - 13 1058 75
AY263368.1 - 15 - 16 1724 75
BC139012.1 - 15 - 16 1655 75
BC145500.1 - 15 - 16 1652 75

View Full Product Details