The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290633.1 | NP_001277562.1 | 17 - 18 | 2026 | 68 |
NM_178256.4 | NP_839987.2 | 17 - 18 | 2029 | 68 | |
GenBank mRNA | AK147418.1 | - | 15 - 16 | 1491 | 68 |
AK147432.1 | - | 14 - 15 | 1308 | 68 | |
AY263368.1 | - | 17 - 18 | 1974 | 68 | |
BC139012.1 | - | 17 - 18 | 1905 | 68 | |
BC145500.1 | - | 17 - 18 | 1902 | 68 |